Anti-CTDSPL Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB300-231B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Synaptonemal Complex Protein 3
- Antigen symbol:SCP3
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Mouse
- Gene ID:50511
- Antigen synonyms:Synaptonemal complex protein 3|RNASCP3|COR1|Sycp3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
The SCP3 / SYCP3 Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to SCP3 / SYCP3. This antibody reacts with human, mouse. The SCP3 / SYCP3 Antibody [Biotin] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:750
Type: Primary
Antigen: SCP3/SYCP3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse