Anti-CTDSPL Rabbit Polyclonal Antibody
Type: Primary
Antigen: CTDSPL (CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) small phosphatase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
- Catalog No:
- NOVUNB300-231
- NOVUNB300-231SS
- Antigen-Symbol:
- SCP3
- SCP3
- Antigen-Name:
- Synaptonemal Complex Protein 3
- Synaptonemal Complex Protein 3
- Antigen-Synonyme:
- Synaptonemal complex protein 3|RNASCP3|COR1|Sycp3
- Synaptonemal complex protein 3|RNASCP3|COR1|Sycp3
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Polyklonal
- Polyklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Mouse
- Mouse
- Wirt:
- Rabbit
- Rabbit
- Gen-ID:
- 50511
- 50511
- Western blot:
- Yes
- Yes
- ImmunoChemie:
- Yes
- Yes
- ImmunFluoreszenz:
- Yes
- Yes
- Lagertemperatur:
- Cool
- Cool
- Transporttemperatur:
- Ice
- Ice
Frequently Bought Together