Anti-SLC5A7 Mouse Monoclonal Antibody [clone: 62-2E8]
Supplier: Novus Biologicals
Certificates
About this item
The CHT1 Antibody (62-2E8) from Novus Biologicals is a mouse monoclonal antibody to CHT1. This antibody reacts with human, mouse, rat, primate. The CHT1 Antibody (62-2E8) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:500
Type: Primary
Antigen: CHT1
Clonality: Monoclonal
Clone: 62-2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Primate
Specifications
- Catalog No:
- NOVUNB110-74570SS
- NOVUNB110-74570
- Antigen symbol:
- CHT1
- CHT1
- Antigen name:
- solute carrier family 5 (sodium/choline cotransporter), member 7
- solute carrier family 5 (sodium/choline cotransporter), member 7
- Antigen synonyms:
- HMN7A|CHT|CHT1
- HMN7A|CHT|CHT1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 62-2E8
- 62-2E8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 60482
- 60482
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together