Anti-CDR2 Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The CDR2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to CDR2. This antibody reacts with human, mouse, bovine, primate. The CDR2 Antibody has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Recommended Dilutions: Immunohistochemistry (Parafin): 5-10 ?g/ml
Type: Primary
Antigen: CDR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Bovine, Primate
Specifications
- Catalog No:
- NOVUNB110-58345SS
- NOVUNB110-58345
- Antigen symbol:
- CDR2
- CDR2
- Antigen name:
- Cerebellar degeneration-related protein 2, 62kDa
- Cerebellar degeneration-related protein 2, 62kDa
- Antigen synonyms:
- CDR62|Yo
- CDR62|Yo
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 1039
- 1039
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage buffer:
- Tris-Citrate/Phosphate (pH 7,0 - 8,0)
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together