Anti-CITED4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HT13-2D6.3]
Catalog # NOVUNB110-41572APC
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4
- Antigen symbol:CITED4
- Clonality:Monoclonal
- Clone:HT13-2D6.3
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:163732
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Type: Primary
Antigen: CITED4 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 4)
Clonality: monoclonal
Clone: HT13-2D6.3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human