Anti-KDR Rabbit Polyclonal Antibody (FITC (Fluorescein))
Katalognr. NOVUNB100-529F
Supplier: Novus Biologicals
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:kinase insert domain receptor
- Antigen-Symbol:KDR
- Klonalität:Polyklonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Wirt:Rabbit
- Reaktivität:Human,Mouse
- Western blot:Yes
- Gen-ID:3791
- Antigen-Synonyme:VEGFR|CD309|VEGFR2|FLK1
- Lagertemperatur:Cool
- Transporttemperatur:Ice
- Größe:100 μL
- VE:0,1 mL
Specifications
About this item
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse