Anti-KDR Rabbit Polyclonal Antibody (DyLight® 650)
NOVUNB100-529C
- Antibody type:Primary
- Antigen name:Kinase insert Domain receptor
- Antigen symbol:KDR
- Clonality:Polyclonal
- Conjugation:DyLight® 650
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:3791
- Antigen synonyms:VEGFR|Kinase insert domain receptor|CD309|VEGFR2|FLK1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: KDR (kinase insert domain receptor (a type III receptor tyrosine kinase))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse