Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-FLT1 Rabbit Polyclonal Antibody (DyLight® 405)
  NOVUNB100-527V
Anti-FLT1 Rabbit Polyclonal Antibody (DyLight® 405)
  NOVUNB100-527V

 

  • Antibody type:
    Primary
  • Antigen name:
    FMS-related tyrosine kinase 1
  • Antigen symbol:
    FLT1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 405
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    2321
  • Antigen synonyms:
    FLT-1|FLT|VEGFR1|VEGFR-1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μL
  • Pk:
    0,1 mL

 

 

Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human