Order Entry
Switzerland
ContactUsLinkComponent
Anti-DERL1 Rabbit Polyclonal Antibody
Anbieter:  Novus Biologicals
Zertifikate
undefined
Anti-DERL1 Rabbit Polyclonal Antibody
Anbieter:  Novus Biologicals

Technische Daten

  • Catalog No:
  • NOVUNB100-448
  • NOVUNB100-448SS
  • Antigen symbol:
  • DERL1
  • DERL1
  • Antigen name:
  • Derlin 1
  • Derlin 1
  • Antigen synonyms:
  • DER-1|DER1|Der1-like Domain Family member 1
  • DER-1|DER1|Der1-like Domain Family member 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 79139
  • 79139
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Technische Daten

Zugehörige Informationen

Frequently Bought Together