Anti-14-3-3 gamma Mouse Monoclonal Antibody [clone: HS23]
Supplier: Novus Biologicals
Certificates
About this item
Type: Primary
Antigen: 14-3-3 gamma (14-3-3 gamma)
Clonality: monoclonal
Clone: HS23
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Specifications
- Catalog No:
- NOVUNB100-407SS
- NOVUNB100-407
- Antigen symbol:
- 14-3-3 gamma
- 14-3-3 gamma
- Antigen name:
- Protein kinase C inhibitor protein 1
- Protein kinase C inhibitor protein 1
- Antigen synonyms:
- KCIP-1
- KCIP-1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- HS23
- HS23
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7532
- 7532
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together