Order Entry
Switzerland
ContactUsLinkComponent
Anti-MRE11A Rabbit Polyclonal Antibody
Anti-MRE11A Rabbit Polyclonal Antibody
Catalog # NOVUNB100-142SS
Supplier:  Novus Biologicals
Anti-MRE11A Rabbit Polyclonal Antibody
Catalog # NOVUNB100-142SS
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Meiotic Recombination 11
  • Antigen symbol:
    MRE11
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    4361
  • Antigen synonyms:
    ATLD|HNGS1|MRE11 homolog A|meiotic recombination 11 homolog A|MRE11B|MRE11|meiotic recombination 11 homolog 1|double-strand break repair protein MRE11A|DNA recombination and repair protein|AT-like disease|MRE11 homolog 1|endo/exonuclease Mre11
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    25 µL
  • Pk:
    0,025 mL

Specifications

About this item

Type: Primary
Antigen: MRE11A (MRE11 meiotic recombination 11 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: