Order Entry
Switzerland
ContactUsLinkComponent
Anti-MIF Rabbit Polyclonal Antibody
Anti-MIF Rabbit Polyclonal Antibody
Catalog # BSBTRP1020
Supplier:  Boster Bio
Anti-MIF Rabbit Polyclonal Antibody
Catalog # BSBTRP1020
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Macrophage Migration Inhibitory Factor
  • Antigen symbol:
    MIF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    MMIF|phenylpyruvate tautomerase|GLIF|L-dopachrome tautomerase|GIF|macrophage migration inhibitory factor|L-dopachrome isomerase
  • Amino acid number:
    1 - 115
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    E. coli-derived human MIF recombinant protein (Position: M1-A115)
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Macrophage migration inhibitory factor(MIF) detection. Tested with WB, ELISA in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: MIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human