Anti-GCLC Rabbit Polyclonal Antibody
Catalog # BSBTPA1492
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Glutamate-cysteine ligase, catalytic subunit
- Antigen symbol:GCLC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:N-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:GCS|gamma-ECS|GCS heavy chain|glutamate--cysteine ligase catalytic subunit|gamma-glutamylcysteine synthetase|GLCL|GLCLC|GCL
- Amino acid number:7 - 24
- Sequence:GSPLSWEETKRHADHVRR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human GCLC (7-24aa GSPLSWEETKRHADHVRR), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Glutamate--cysteine ligase catalytic subunit(GCLC) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: GCLC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat