Anti-TIMP3 Rabbit Polyclonal Antibody
Catalog # BSBTPA1077
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Tissue inhibitor of metalloproteinases 3
- Antigen symbol:TIMP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:K222TA2|tissue inhibitor of metalloproteinases 3|TIMP-3|MIG-5 protein|K222|HSMRK222|metalloproteinase inhibitor 3|protein MIG-5|SFD
- Amino acid number:198 - 211
- Sequence:WAPPDKSIINATDP
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human TIMP3 (198-211aa WAPPDKSIINATDP), different from the related rat and mouse sequences by one amino acid
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Metalloproteinase inhibitor 3 (TIMP3) detection. Tested with WB, ELISA in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: TIMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat