Anti-MAT2B/TGR Goat Polyclonal Antibody
Catalog # ABCAAB139132-200
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Methionine adenosyltransferase 2 beta
- Antigen symbol:MAT2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pack type:Vial
- Pk:200 µl
Specifications
About this item
Goat polyclonal to MAT2B/TGR.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MAT2B/TGR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human