Order Entry
Switzerland
ContactUsLinkComponent
Anti-Viperin Mouse Monoclonal Antibody [clone: MaP.VIP]
Anti-Viperin Mouse Monoclonal Antibody [clone: MaP.VIP]
Catalog # ABCAAB107359-50
Supplier:  Abcam
undefined
Anti-Viperin Mouse Monoclonal Antibody [clone: MaP.VIP]
Catalog # ABCAAB107359-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    radical S-adenosyl methionine domain containing 2
  • Antigen symbol:
    RSAD2
  • Clonality:
    Monoclonal
  • Clone:
    MaP.VIP
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal [MaP.VIP] to Viperin.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Viperin
Clonality: Monoclonal
Clone: MaP.VIP
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human;Mouse