Anti-Hsp70 Mouse Monoclonal Antibody [clone: C92F3A-5]
ABCAAB47455-100
: Abcam
- Antibody type:Primary
- Antigen name:Heat shock protein 70kDa family, member 13
- Antigen symbol:HSPA13
- Clonality:Monoclonal
- Clone:C92F3A-5
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pack type:Vial
- Pk:100 µl
Mouse monoclonal [C92F3A-5] to Hsp70.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Hsp70
Clonality: Monoclonal
Clone: C92F3A-5
Conjugation:
Epitope: The mapped epitope is in the region of amino acid residues 436-503.
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat