Order Entry
Switzerland
ContactUsLinkComponent
Anti-HIV2 gp36 Mouse Monoclonal Antibody [clone: 3B10/G2]
Anti-HIV2 gp36 Mouse Monoclonal Antibody [clone: 3B10/G2]
Catalog # ABCAAB18628-100
Supplier:  Abcam
Anti-HIV2 gp36 Mouse Monoclonal Antibody [clone: 3B10/G2]
Catalog # ABCAAB18628-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HIV2 gp36
  • Clonality:
    Monoclonal
  • Clone:
    3B10/G2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [3B10/G2] to HIV2 gp36.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIV2 gp36
Clonality: Monoclonal
Clone: 3B10/G2
Conjugation:
Epitope: Recognition epitope resides between amino acid residues 390a.a.-702 a.a. of HIV gp36 antigen.
Host: Mouse
Isotype: IgG2a
Reactivity: HIV2