Order Entry
Switzerland
ContactUsLinkComponent
 
 
Anti-Ganglioside GD3 Mouse Monoclonal Antibody [clone: R24]
  ABCAAB11779-500
 :  Abcam
undefined
Anti-Ganglioside GD3 Mouse Monoclonal Antibody [clone: R24]
  ABCAAB11779-500
 :  Abcam

 

  • Antibody type:
    Primary
  • Antigen name:
    Ganglioside GD3
  • Clonality:
    Monoclonal
  • Clone:
    R24
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Seq:
    1418
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µl
  • Pack type:
    Vial
  • Pk:
    500 µl

 

 

Mouse monoclonal [R24] to Ganglioside GD3.

 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Ganglioside GD3
Clonality: Monoclonal
Clone: R24
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human