Order Entry
Switzerland
ContactUsLinkComponent
Anti-Cyclin B2/CCNB2 Mouse Monoclonal Antibody [clone: X121.10]
Anti-Cyclin B2/CCNB2 Mouse Monoclonal Antibody [clone: X121.10]
Catalog # ABCAAB10839-50
Supplier:  Abcam
undefined
Anti-Cyclin B2/CCNB2 Mouse Monoclonal Antibody [clone: X121.10]
Catalog # ABCAAB10839-50
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cyclin B2
  • Antigen symbol:
    CCNB2
  • Clonality:
    Monoclonal
  • Clone:
    X121.10
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Xenopus laevis
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pack type:
    Vial
  • Pk:
    50 µG

Specifications

About this item

Mouse monoclonal [X121.10] to Cyclin B2/CCNB2.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cyclin B2/CCNB2
Clonality: Monoclonal
Clone: X121.10
Conjugation:
Epitope: The epitope for X121 lies within a region spanning the amino acids 1-112
Host: Mouse
Isotype: IgG1
Reactivity: Xenopus laevis