Anti-SERCA1 ATPase Mouse Monoclonal Antibody [clone: IIH11]
Supplier: Abcam
New Product
Certificates
About this item
Mouse monoclonal [IIH11] to SERCA1 ATPase.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SERCA1 ATPase
Clonality: Monoclonal
Clone: IIH11
Conjugation:
Epitope: This antibody recognizes an epitope between amino acids 199-505 of rabbit skeletal muscle ATPase, a region that is not exposed in native sarcoplasmic reticulum
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat
Specifications
- Catalog No:
- ABCAAB2818-100
- ABCAAB2818-1MG
- Antigen name:
- SERCA1 ATPase
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- IIH11
- Host:
- Mouse
- Isotype:
- IgG1
Specifications
Frequently Bought Together