Anti-Hepatitis C Virus Core Antigen Mouse Monoclonal Antibody [clone: 1F6] (Biotinylated)
Catalog # ABCAAB2587-50
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Hepatitis C Virus Core Antigen
- Clonality:Monoclonal
- Clone:1F6
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Seq:1202
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pack type:Vial
- Pk:50 µG
Specifications
About this item
Biotin Mouse monoclonal [1F6] to Hepatitis C Virus Core Antigen.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Hepatitis C Virus Core Antigen
Clonality: Monoclonal
Clone: 1F6
Conjugation: Biotin
Epitope: This antibody recognises amino acid residues 80-120 of Hepatitis C Core Antigen.
Host: Mouse
Isotype: IgG2a
Reactivity: Hepatitis C virus (HCV)