Anti-Fibronectin Mouse monoclonal antibody unconjugated
BIRBORB95640-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Fibronectin
- Antigen symbol:FN1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Storage buffer:HEPES with 0,15M NaCl, 0,01% BSA, 0,03% sodium azide, and 50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by ammonium sulfate precipitation
- Size:50 µg
- Pk:50 µG
Immunochemistry: 1:200: 1:2000
Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human