Anti-SLC5A6 Rabbit Polyclonal Antibody
Catalog # BIRBORB87856-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
- Antigen symbol:Solute carrier family 5, member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:PBS, 0,1% sodium azide.
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Specifications
About this item
- Immunochemistry: 1.25-2.5 μg/ml
Type: Primary
Antigen: SLC5A6 (solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human