Auftragserfassung
Switzerland
ContactUsLinkComponent
Anti-SLC5A6 Rabbit Polyclonal Antibody
Anti-SLC5A6 Rabbit Polyclonal Antibody
Catalog # BIRBORB87768-50
Supplier:  Biorbyt
undefined
Anti-SLC5A6 Rabbit Polyclonal Antibody
Catalog # BIRBORB87768-50
Supplier:  Biorbyt

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
  • Antigen-Symbol:
    Solute carrier family 5, member 6
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Reaktivität:
    Human
  • Lagerungspuffer:
    PBS, 0,1% sodium azide.
  • Lagertemperatur:
    Cool
  • Konzentration:
    1 mg/ml
  • Transporttemperatur:
    Ice
  • Reinigung:
    purified by immunoaffinity chromatography
  • Größe:
    50 µg
  • VE:
    50 µG

Specifications

About this item

  • Immunochemistry: 10 μg/ml

Type: Primary
Antigen: SLC5A6 (solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human