Anti-AMPK2 Rabbit Polyclonal Antibody
Catalog # BIRBORB77065-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 2 catalytic subunit
- Antigen symbol:PRKAA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:AMPK alpha 2|PRKAA2
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Peptide
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:20000-1:80000; Western Blot: 1:200-1:1000
Type: Primary
Antigen: AMPK2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat