Order Entry
Switzerland
ContactUsLinkComponent
Anti-SLC12A3 Rabbit Polyclonal Antibody (Cy5.5®)
Anti-SLC12A3 Rabbit Polyclonal Antibody (Cy5.5®)
  BIRBORB123380-100
 :  Biorbyt
Anti-SLC12A3 Rabbit Polyclonal Antibody (Cy5.5®)
  BIRBORB123380-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 12 (sodium/chloride transporter), member 3
  • Antigen symbol:
    Solute carrier family 12, member A3
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5.5®
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Form:
    liquid
  • Storage buffer:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Molecular weight:
    110 - 160 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: SLC12A3 (solute carrier family 12 (sodium/chloride transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat