Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-PRKAA1/A2 Rabbit Polyclonal Antibody
  BIRBORB109129-10
 :  Biorbyt
undefined
Anti-PRKAA1/A2 Rabbit Polyclonal Antibody
  BIRBORB109129-10
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    protein kinase, AMP-activated, alpha 1/2
  • Antigen symbol:
    PRKAA1/A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Modification:
    phospho-T172
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Chemically synthesized phosphopeptide derived from the region of human AMPK that contains threonine 172. The sequence is conserved in human, mouse and rat.
  • Purification:
    purified by antigen immunoaffinity chromatography
  • Size:
    10 Tests
  • Pk:
    10

 

 

Type: Primary
Antigen: PRKAA1/A2 (protein kinase, AMP-activated, alpha 1/2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat