Anti-PRKAA1 Rabbit Polyclonal Antibody
BIRBORB106235-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Modification:phospho-Ser485
- Storage buffer:Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:62 kDa
- Storage temperature:Cool
- Concentration:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Shipping temperature:Ice
- Immunogen:A synthesized peptide derived from human AMPK1 around the phosphorylation site of Serine 485
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat