Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-Methamphetamine (M-AMP) Mouse Monoclonal Antibody
  BIRBORB24518-1
 :  Biorbyt
Anti-Methamphetamine (M-AMP) Mouse Monoclonal Antibody
  BIRBORB24518-1
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Methamphetamine
  • Antigen symbol:
    M-AMP
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Storage buffer:
    PBS, pH 7.0
  • Storage temperature:
    Cool
  • Concentration:
    4,5 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    Methamphetamine-BSA conjugate
  • Size:
    1 mg
  • Pk:
    1 mg

 

 

Type: Primary
Antigen: Methamphetamine (M-AMP)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: