Anti-PRKAB1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Katalognr. BIRBORB15085-100
Anbieter: Biorbyt
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Protein Kinase AMP-Activated Non-Catalytic Subunit Beta 1
- Antigen-Symbol:PRKAB1
- Klonalität:Polyklonal
- Konjugation:FITC (Fluorescein Isothiocyanate)
- Wirt:Rabbit
- ImmunFluoreszenz:Yes
- Isotyp:IgG
- Reaktivität:
- Environmentally Preferable:
- Form:liquid
- Lagerungspuffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:30 kDa
- Lagertemperatur:Cool
- Konzentration:0,5 mg/ml
- Transporttemperatur:Ice
- Immunogen:Synthetic peptide
- Reinigung:purified by affinity chromatography
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: PRKAB1 (protein kinase, AMP-activated, beta 1 non-catalytic subunit)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: