Anti-NRBP2 Mouse Monoclonal Antibody [clone: 4D1]
Catalog # ABNOH00340371-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Binding Protein 2
- Antigen symbol:NRBP2
- Clonality:Monoclonal
- Clone:4D1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- ChIP:Yes
- Gene ID:340371
- Antigen synonyms:BC011468
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NRBP2
Clonality: Monoclonal
Clone: 4D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human