Anti-DUSP15 Rabbit Polyclonal Antibody
100 µg
Catalog # ABNOH00128853-D01P
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: DUSP15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00128853-D01P.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Dual specificity phosphatase 15
- Cat. No.:ABNOH00128853-D01P
- Reactivity:Human
- Gene ID:128853
- Antigen synonyms:FLJ40111|VHY|RP11-243J16.11
- Conjugation:Unconjugated
- Antigen symbol:DUSP15



