Anti-CERS4 Mouse Monoclonal Antibody [clone: 4B10]
Catalog # ABNOH00079603-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ceramide synthase 4
- Antigen symbol:CERS4
- Clonality:Monoclonal
- Clone:4B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:79603
- Antigen synonyms:Trh1|LASS4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CERS4
Clonality: Monoclonal
Clone: 4B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human