Anti-CXCR6 Rabbit Polyclonal Antibody
Catalog # ABNOH00010663-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) Receptor 6
- Antigen symbol:CXCR6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10663
- Antigen synonyms:TYMSTR|C-X-C chemokine receptor type 6|CD186|BONZO|STRL33|Bonzo|CDw186|CXC-R6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CXCR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human