Anti-UBTF Mouse Monoclonal Antibody [clone: 6C6]
Catalog # ABNOH00007343-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Upstream binding transcription factor, RNA polymerase I
- Antigen symbol:UBTF
- Clonality:Monoclonal
- Clone:6C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:7343
- Antigen synonyms:UBF-1|Upstream-binding factor 1|Nucleolar transcription factor 1|UBF1|Autoantigen NOR-90|UBF|UBTF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UBTF
Clonality: Monoclonal
Clone: 6C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat