Anti-TSG101 Mouse Monoclonal Antibody [clone: 5B7]
ABNOH00007251-M01
: Abnova
- Antibody type:Primary
- Antigen name:Tumor Susceptibility 101
- Antigen symbol:TSG101
- Clonality:Monoclonal
- Clone:5B7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- ChIP:Yes
- Gene ID:7251
- Antigen synonyms:VPS23|TSG10|Tumor susceptibility gene 101 protein
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TSG101
Clonality: Monoclonal
Clone: 5B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human