Anti-PRKAB1 Mouse Monoclonal Antibody [clone: 1D3]
Catalog # ABNOH00005564-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein Kinase, AMP-activated Non-catalytic Subunit Beta 1
- Antigen symbol:PRKAB1
- Clonality:Monoclonal
- Clone:1D3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5564
- Antigen synonyms:kinase AMPK-beta1|AMPK-beta1|AMPK beta-1 chain|5'-AMP-activated protein kinase beta-1 subunit|PRKAB1|AAKB1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PRKAB1
Clonality: Monoclonal
Clone: 1D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse