Anti-IRF3 Rabbit Polyclonal Antibody
Catalog # ABNOH00003661-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Interferon regulatory factor 3
- Antigen symbol:IRF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3661
- Antigen synonyms:Interferon regulatory factor 3|IRF3|IRF-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: IRF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human