Anti-IMPDH2 Rabbit Polyclonal Antibody
ABNOH00003615-D01P
: Abnova
- Antibody type:Primary
- Antigen name:Inosine-5'-monophosphate Dehydrogenase 2
- Antigen symbol:IMPDH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:3615
- Antigen synonyms:inosine 5' phosphate dehydrogenase 2|IMPD2|inosine monophosphate dehydrogenase type II|IMP oxireductase 2|inosine monophosphate oxireductase 2|IMPDH2|IMP dehydrogenase 2|inosine monophosphate dehydrogenase 2|IMP (inosine monophosphate) dehydrogenase 2|IMPDH-II
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: IMPDH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human