Order Entry
Switzerland
ContactUsLinkComponent
Anti-CLCN7 Mouse Monoclonal Antibody [clone: 4A3]
Anti-CLCN7 Mouse Monoclonal Antibody [clone: 4A3]
  ABNOH00001186-M01A
 :  Abnova
undefined
Anti-CLCN7 Mouse Monoclonal Antibody [clone: 4A3]
  ABNOH00001186-M01A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    chloride channel, voltage-sensitive 7
  • Antigen symbol:
    CLCN7
  • Clonality:
    Monoclonal
  • Clone:
    4A3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1186
  • Antigen synonyms:
    CLC7|OPTB4|OPTA2|CLC-7|PPP1R63
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: CLCN7
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human