Anti-SLC5A6 Rabbit Polyclonal Antibody
Catalog # ORIGTA314828
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6
- Antigen symbol:Solute carrier family 5, member 6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
- Recommended Dilutions: Western Blot: 1:500-1:3000; ELISA: 1:10000
Type: Primary
Antigen: SLC5A6 (solute carrier family 5 (sodium/multivitamin and iodide cotransporter), member 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human