Anti-PRKAA1 Rabbit Monoclonal Antibody [clone: Y365]
Catalog # ORIGTA303654
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Protein kinase, AMP-activated, alpha 1 catalytic subunit
- Antigen symbol:PRKAA1
- Clonality:Monoclonal
- Clone:Y365
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS 49%,Sodium azide 0,01%,Glycerol 50%,BSA 0,05%
- Molecular weight:63 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Flow Cytometry: 1:100; Western Blot: 1:1000-1:5000; Immunochemistry: 1:100-1:250; Immunoprecipitation: 1:50
Type: Primary
Antigen: PRKAA1 (protein kinase, AMP-activated, alpha 1 catalytic subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat