Mouse IgG1 Isotype Control [clone: MOPC-21] (PE (Phycoerythrin)/Cy7®)
60-4714-U100
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:PE (Phycoerythrin)/Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: