Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-CD8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RPA-T8]
 
undefined
Anti-CD8A Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RPA-T8]

 

  • Catalog No:
  • 20-0088-T025
  • 20-0088-T500
  • 20-0088-T100
  • Antigen symbol:
  • CD8A
  • CD8A
  • CD8A
  • Antigen name:
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • Antigen synonyms:
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • RPA-T8
  • RPA-T8
  • RPA-T8
  • Reactivity:
  • Baboon, Cynomolgus monkey, Sooty mangabey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Sooty mangabey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Sooty mangabey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • 925
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Ice

 

Frequently Bought Together