Anti-NSMCE4A Rabbit Polyclonal Antibody
ABGEAP9909A
: Abgent
- Antibody type:Primary
- Antigen name:Non-SMC element 4 homolog A (S. cerevisiae)
- Antigen symbol:NSMCE4A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal
- Gene ID:10365
- Antigen synonyms:LKLF|KLF2
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequence:PRAEPESGGTDDDLNC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,4 mg/ml
- Shipping temperature:4 °C
- Immunogen:This KLF2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 13-42 amino acids from the N-terminal region of human KLF2.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: NSMCE4A (non-SMC element 4 homolog A (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human