Anti-SLC5A7 Rabbit Polyclonal Antibody
Catalog # AVIVARP44024_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 5 (sodium/choline cotransporter), member 7
- Antigen symbol:Solute carrier family 5, member 7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:60482
- Antigen synonyms:HMN7A|CHT|CHT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SLC5A7 (solute carrier family 5 (sodium/choline cotransporter), member 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: