Order Entry
Switzerland
ContactUsLinkComponent
Anti-SLC15A2 Rabbit Polyclonal Antibody
Anti-SLC15A2 Rabbit Polyclonal Antibody
  AVIVARP44016_P050
Anti-SLC15A2 Rabbit Polyclonal Antibody
  AVIVARP44016_P050

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 15 (oligopeptide transporter), member 2
  • Antigen symbol:
    Solute carrier family 15, member 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Epitope:
    middle
  • Cross adsorption:
    none
  • Gene ID:
    6565
  • Antigen synonyms:
    PEPT2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: SLC15A2 (solute carrier family 15 (oligopeptide transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: