Anti-ARPP21 Rabbit Polyclonal Antibody
Catalog # AVIVARP41873_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:cyclic AMP-regulated phosphoprotein, 21
- Antigen symbol:ARPP21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:74100
- Antigen synonyms:R3hdm3|Tarpp|0710001E13Rik|AI853636|ARPP-21|D9Bwg1012e
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: ARPP21 (cAMP-regulated phosphoprotein, 21kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: