Anti-HOXB9 Rabbit Polyclonal Antibody
Catalog # AVIVARP39275_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:homeobox B9
- Antigen symbol:HOXB9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:15417
- Antigen synonyms:Hox-2.5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HOXB9 (homeobox B9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: